Assay Details
| Gene Symbol: LOC419677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947881.3 | XP_004947938.1 | 8 - 9 | 1863 | 70 | |
| XM_004947882.2 | XP_004947939.1 | 10 - 11 | 1074 | 70 | ||
| XM_004947884.3 | XP_004947941.1 | 10 - 11 | 1901 | 70 | ||
| XM_015297911.2 | XP_015153397.1 | 9 - 10 | 866 | 70 | ||
| XM_015297913.2 | XP_015153399.1 | 9 - 10 | 868 | 70 | ||
| XM_025143103.1 | XP_024998871.1 | 8 - 9 | 1248 | 70 | ||
| XM_025143104.1 | XP_024998872.1 | 9 - 10 | 993 | 70 | ||
| XM_025143105.1 | XP_024998873.1 | 11 - 12 | 1139 | 70 | ||
| XM_025143106.1 | XP_024998874.1 | 11 - 12 | 1981 | 70 | ||
| XM_025143107.1 | XP_024998875.1 | 9 - 10 | 866 | 70 | ||
| GenBank mRNA | CR386316.1 | - | 4 - 5 | 604 | 70 | |
More Information
Set Membership: |
|

LOC419677
Genomic Map