Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291703.2 | XP_015147189.2 | 13 - 14 | 2368 | 92 | |
| GenBank mRNA | CR386338.1 | - | 2 - 3 | 274 | 92 | |
More Information
Set Membership: |
|

NAALADL2
Genomic Map