Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155078.1 | XP_025010846.1 | 10 | 3244 | 86 | |
| XM_025155079.1 | XP_025010847.1 | 10 | 3241 | 86 | ||
| XM_025155082.1 | XP_025010850.1 | 10 | 3062 | 86 | ||
| XM_025155085.1 | XP_025010853.1 | 10 | 3062 | 86 | ||
| GenBank mRNA | CR386585.1 | - | 1 | 820 | 86 | |
More Information
Set Membership: |
|

SEPTIN3
Genomic Map