Assay Details
| Gene Symbol: GMCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142818.1 | XP_024998586.1 | 5 - 6 | 749 | 94 | |
| XM_025142819.1 | XP_024998587.1 | 4 - 5 | 609 | 94 | ||
| GenBank mRNA | CR386694.1 | - | 4 - 5 | 397 | 94 | |
More Information
Set Membership: |
|

GMCL1
Genomic Map