Assay Details
| Gene Symbol: SAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273056.2 | XP_015128542.1 | 6 - 7 | 977 | 55 | |
| XM_015273057.2 | XP_015128543.1 | 6 - 7 | 855 | 55 | ||
| XM_025144511.1 | XP_025000279.1 | 6 - 7 | 981 | 55 | ||
| GenBank mRNA | CR386719.1 | - | 6 - 7 | 751 | 55 | |
More Information
Set Membership: |
|

SAE1
Genomic Map