Assay Details
| Gene Symbol: CCBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277496.2 | XP_015132982.1 | 11 | 4687 | 77 | |
| GenBank mRNA | CR386734.1 | - | 1 | 822 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCBE1| Gene Symbol: CCBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277496.2 | XP_015132982.1 | 11 | 4687 | 77 | |
| GenBank mRNA | CR386734.1 | - | 1 | 822 | 77 | |
Set Membership: |
|