Assay Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936279.3 | XP_004936336.2 | 3 - 4 | 917 | 98 | |
| XM_025149859.1 | XP_025005627.1 | 2 - 3 | 422 | 98 | ||
| XM_420873.6 | XP_420873.3 | 3 - 4 | 1028 | 98 | ||
| GenBank mRNA | CR386840.1 | - | 2 - 3 | 583 | 98 | |
More Information
Set Membership: |
|

PANK2
Genomic Map