Assay Details
| Gene Symbol: TYW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142014.1 | XP_024997782.1 | 16 | 3542 | 95 | |
| GenBank mRNA | CR387006.1 | - | 1 | 1124 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TYW1| Gene Symbol: TYW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142014.1 | XP_024997782.1 | 16 | 3542 | 95 | |
| GenBank mRNA | CR387006.1 | - | 1 | 1124 | 95 | |
Set Membership: |
|