Assay Details
| Gene Symbol: PNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299479.1 | XP_015154965.1 | 5 - 6 | 528 | 57 | |
| GenBank mRNA | CR387260.1 | - | 5 - 6 | 509 | 57 | |
| CR524459.1 | - | 5 - 6 | 499 | 57 | ||
More Information
Set Membership: |
|

PNPO
Genomic Map