Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293494.2 | XP_015148980.1 | 3 - 4 | 592 | 104 | |
| GenBank mRNA | CR387521.1 | - | 2 - 3 | 331 | 104 | |
More Information
Set Membership: |
|

LSMEM2
Genomic Map