Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295908.1 | XP_015151394.1 | 7 - 8 | 1063 | 84 | |
| GenBank mRNA | CR387663.1 | - | 3 - 4 | 332 | 84 | |
More Information
Set Membership: |
|

POM121C
Genomic Map