Assay Details
| Gene Symbol: SEH1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282472.2 | XP_015137958.1 | 9 | 2084 | 78 | |
| GenBank mRNA | CR388472.1 | - | 1 | 622 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEH1L| Gene Symbol: SEH1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282472.2 | XP_015137958.1 | 9 | 2084 | 78 | |
| GenBank mRNA | CR388472.1 | - | 1 | 622 | 78 | |
Set Membership: |
|