Assay Details
| Gene Symbol: GLIS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642989.4 | XP_003643037.2 | 4 - 5 | 3802 | 61 | |
| XM_004949062.2 | XP_004949119.1 | 4 - 5 | 1748 | 61 | ||
| GenBank mRNA | CR389088.1 | - | 2 - 3 | 274 | 61 | |
More Information
Set Membership: |
|

GLIS3
Genomic Map