Assay Details
| Gene Symbol: SMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR389146.1 | - | 3 - 4 | 417 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMPD3| Gene Symbol: SMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR389146.1 | - | 3 - 4 | 417 | 104 | |
Set Membership: |
|