Assay Details
| Gene Symbol: STPG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945781.3 | XP_004945838.1 | 2 - 3 | 485 | 69 | |
| XM_415538.5 | XP_415538.1 | 2 - 3 | 485 | 69 | ||
| GenBank mRNA | CR389219.1 | - | 2 - 3 | 144 | 69 | |
More Information
Set Membership: |
|

STPG3
Genomic Map