Assay Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942058.3 | XP_004942115.2 | 6 | 10395 | 85 | |
| GenBank mRNA | CR389248.1 | - | 1 | 626 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IPMK| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942058.3 | XP_004942115.2 | 6 | 10395 | 85 | |
| GenBank mRNA | CR389248.1 | - | 1 | 626 | 85 | |
Set Membership: |
|