Assay Details
| Gene Symbol: ABCB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284389.2 | XP_015139875.1 | 10 - 11 | 1795 | 95 | |
| GenBank mRNA | CR389335.1 | - | 1 - 2 | 183 | 95 | |
| CR406644.1 | - | 1 - 2 | 131 | 95 | ||
More Information
Set Membership: |
|

ABCB10
Genomic Map