Assay Details
| Gene Symbol: CNNM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003643431.4 | XP_003643479.3 | 1 - 2 | 1469 | 93 | |
| GenBank mRNA | CR389457.1 | - | 1 - 2 | 558 | 93 | |
More Information
Set Membership: |
|

CNNM4
Genomic Map