Assay Details
| Gene Symbol: PARP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292850.2 | XP_015148336.1 | 10 - 11 | 79 | ||
| XM_015292851.2 | XP_015148337.1 | 10 - 11 | 79 | |||
| XM_015292852.2 | XP_015148338.1 | 10 - 11 | 79 | |||
| XM_025154688.1 | XP_025010456.1 | 9 - 10 | 79 | |||
| GenBank mRNA | CR389687.1 | - | 7 - 8 | 996 | 79 | |
More Information
Set Membership: |
|

PARP3
Genomic Map