Assay Details
| Gene Symbol: C28H19ORF44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948936.3 | XP_004948993.1 | 1 - 2 | 60 | 79 | |
| XM_418261.6 | XP_418261.4 | 1 - 2 | 132 | 79 | ||
| GenBank mRNA | CR389710.1 | - | 1 - 2 | 60 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004948936.3, XM_418261.6 transcripts.
Set Membership: |
|

C28H19ORF44
Genomic Map