Assay Details
| Gene Symbol: BNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943823.3 | XP_004943880.1 | 5 | 3842 | 115 | |
| XM_425076.6 | XP_425076.2 | 5 | 4195 | 115 | ||
| GenBank mRNA | CR389812.1 | - | 1 | 325 | 116 | |
More Information
Set Membership: |
|

BNC1
Genomic Map