Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283985.2 | XP_015139471.1 | 3 | 3468 | 152 | |
| XM_015283986.2 | XP_015139472.1 | 2 | 3384 | 152 | ||
| GenBank mRNA | CR390023.1 | - | 1 | 216 | 152 | |
| CR390411.1 | - | 1 | 214 | 152 | ||
More Information
Set Membership: |
|

ZBTB18
Genomic Map