Assay Details
| Gene Symbol: RBM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274638.2 | XP_015130124.1 | 4 - 5 | 437 | 107 | |
| GenBank mRNA | CR390112.1 | - | 4 - 5 | 340 | 107 | |
More Information
Set Membership: |
|

RBM8A
Genomic Map