Assay Details
| Gene Symbol: ALG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298312.2 | XP_015153798.1 | 6 - 7 | 4423 | 72 | |
| XM_025143254.1 | XP_024999022.1 | 6 - 7 | 4423 | 72 | ||
| XM_025143255.1 | XP_024999023.1 | 2 - 3 | 191 | 72 | ||
| GenBank mRNA | CR390311.1 | - | 2 - 3 | 161 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025143255.1 transcripts.
Set Membership: |
|

ALG9
Genomic Map