Assay Details
| Gene Symbol: VPS4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292679.2 | XP_015148165.2 | 6 - 7 | 696 | 93 | |
| GenBank mRNA | CR390723.1 | - | 1 - 2 | 336 | 93 | |
More Information
Set Membership: |
|

VPS4A
Genomic Map