Assay Details
| Gene Symbol: LOC101749023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278810.2 | XP_015134296.2 | 13 | 4154 | 70 | |
| XM_015278817.2 | XP_015134303.2 | 13 | 4220 | 70 | ||
| XM_015278822.2 | XP_015134308.2 | 13 | 4861 | 70 | ||
| XM_015278832.2 | XP_015134318.2 | 12 | 3996 | 70 | ||
| XM_015278845.2 | XP_015134331.2 | 10 | 3724 | 70 | ||
| XM_015278851.2 | XP_015134337.2 | 11 | 3826 | 70 | ||
| XM_015278861.2 | XP_015134347.2 | 10 | 3637 | 70 | ||
| XM_015278875.2 | XP_015134361.2 | 10 | 3649 | 70 | ||
| XM_025146839.1 | XP_025002607.1 | 12 | 3984 | 70 | ||
| GenBank mRNA | CR390797.1 | - | 1 | 712 | 70 | |
More Information
Set Membership: |
|

LOC101749023
Genomic Map