Assay Details
| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142446.1 | XP_024998214.1 | 3 | 3983 | 80 | |
| GenBank mRNA | CR390913.1 | - | 1 | 322 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STK35| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142446.1 | XP_024998214.1 | 3 | 3983 | 80 | |
| GenBank mRNA | CR390913.1 | - | 1 | 322 | 80 | |
Set Membership: |
|