Assay Details
| Gene Symbol: GIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233884.5 | XP_001233885.3 | 2 - 3 | 325 | 75 | |
| GenBank mRNA | AJ719870.1 | - | 2 - 3 | 325 | 75 | |
More Information
Set Membership: |
|

GIF
Genomic Map