Assay Details
| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642382.4 | XP_003642430.1 | 1 | 639 | 83 | |
| GenBank mRNA | AY435420.1 | - | 1 | 518 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003642382.4 transcripts.
Set Membership: |
|

CLDN4
Genomic Map