Assay Details
| Gene Symbol: LGI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142872.1 | XP_024998640.1 | 6 - 7 | 812 | 59 | |
| GenBank mRNA | AY617124.1 | - | 6 - 7 | 500 | 59 | |
| CR523385.1 | - | 6 - 7 | 500 | 59 | ||
More Information
Set Membership: |
|

LGI3
Genomic Map