Assay Details
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300096.2 | XP_015155582.2 | 1 - 2 | 676 | 69 | |
| GenBank mRNA | AY702439.1 | - | 1 - 2 | 149 | 69 | |
More Information
Set Membership: |
|

NR2F6
Genomic Map