Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934444.3 | XP_004934501.1 | 3 | 2200 | 66 | |
| XM_004934445.3 | XP_004934502.1 | 3 | 2270 | 66 | ||
| XM_025155687.1 | XP_025011455.1 | 3 | 2683 | 66 | ||
| XM_025155688.1 | XP_025011456.1 | 3 | 2325 | 66 | ||
| XM_415296.6 | XP_415296.2 | 3 | 2455 | 66 | ||
| GenBank mRNA | CR391704.1 | - | 1 | 518 | 66 | |
More Information
Set Membership: |
|

GAL3ST1
Genomic Map