Assay Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421754.6 | XP_421754.2 | 4 | 2201 | 66 | |
| GenBank mRNA | CR405865.1 | - | 1 | 714 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP5| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421754.6 | XP_421754.2 | 4 | 2201 | 66 | |
| GenBank mRNA | CR405865.1 | - | 1 | 714 | 66 | |
Set Membership: |
|