Assay Details
| Gene Symbol: CSRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939122.3 | XP_004939179.1 | 5 | 7226 | 70 | |
| XM_418530.6 | XP_418530.3 | 5 | 3944 | 70 | ||
| GenBank mRNA | CR405912.1 | - | 1 | 1214 | 70 | |
More Information
Set Membership: |
|

CSRNP1
Genomic Map