Assay Details
| Gene Symbol: LOC100857911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR406002.1 | - | 1 | 1120 | 67 | |
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154862.1 | XP_025010630.1 | 10 | 7391 | 67 | |
| XM_025154863.1 | XP_025010631.1 | 11 | 7463 | 67 | ||
More Information
Set Membership: |
|

LOC100857911
Genomic Map