Assay Details
| Gene Symbol: MED10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419024.6 | XP_419024.5 | 2 - 3 | 253 | 82 | |
| GenBank mRNA | BX929905.1 | - | 2 - 3 | 214 | 82 | |
| BX933968.2 | - | 2 - 3 | 221 | 82 | ||
| BX934694.2 | - | 2 - 3 | 210 | 82 | ||
More Information
Set Membership: |
|

MED10
Genomic Map