Assay Details
| Gene Symbol: LOC100857622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274577.2 | XP_015130063.1 | 2 - 3 | 386 | 74 | |
| GenBank mRNA | BX930073.2 | - | 2 - 3 | 375 | 74 | |
| BX931946.2 | - | 2 - 3 | 377 | 74 | ||
| BX950608.1 | - | 2 - 3 | 408 | 74 | ||
More Information
Set Membership: |
|

LOC100857622
Genomic Map