Assay Details
| Gene Symbol: LOC100857622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274577.2 | XP_015130063.1 | 4 - 5 | 556 | 57 | |
| GenBank mRNA | BX930073.2 | - | 4 - 5 | 545 | 57 | |
| BX931946.2 | - | 4 - 5 | 547 | 57 | ||
| BX950608.1 | - | 4 - 5 | 578 | 57 | ||
More Information
Set Membership: |
|

LOC100857622
Genomic Map