Assay Details
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296120.2 | XP_015151606.1 | 3 - 4 | 500 | 75 | |
XM_025142002.1 | XP_024997770.1 | 2 - 3 | 418 | 75 | ||
GenBank mRNA | BX930087.1 | - | 2 - 3 | 352 | 75 | |
BX935763.2 | - | 1 - 2 | 164 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
