Assay Details
| Gene Symbol: CCNI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234829.5 | XP_001234830.1 | 4 - 5 | 476 | 74 | |
| XM_004945017.3 | XP_004945074.1 | 4 - 5 | 479 | 74 | ||
| GenBank mRNA | BX930233.2 | - | 4 - 5 | 427 | 74 | |
More Information
Set Membership: |
|

CCNI2
Genomic Map