Assay Details
| Gene Symbol: DUSP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642664.4 | XP_003642712.3 | 1 - 2 | 161 | 117 | |
| GenBank mRNA | BX930262.1 | - | 1 - 2 | 135 | 117 | |
| BX931529.2 | - | 1 - 2 | 254 | 117 | ||
| BX932685.1 | - | 1 - 2 | 117 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003642664.4 transcripts.
Set Membership: |
|

DUSP23
Genomic Map