Assay Details
| Gene Symbol: ARHGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941589.3 | XP_004941646.1 | 6 - 7 | 782 | 87 | |
| XM_004941590.3 | XP_004941647.1 | 5 - 6 | 525 | 87 | ||
| XM_004941591.3 | XP_004941648.1 | 5 - 6 | 934 | 87 | ||
| XM_004941592.3 | XP_004941649.1 | 5 - 6 | 629 | 87 | ||
| XM_015287238.2 | XP_015142724.1 | 7 - 8 | 736 | 87 | ||
| XM_015287239.2 | XP_015142725.1 | 7 - 8 | 703 | 87 | ||
| XM_015287240.1 | XP_015142726.1 | 6 - 7 | 654 | 87 | ||
| XM_015287241.1 | XP_015142727.1 | 6 - 7 | 708 | 87 | ||
| GenBank mRNA | BX930287.1 | - | 5 - 6 | 530 | 87 | |
| BX933100.1 | - | 5 - 6 | 714 | 87 | ||
More Information
Set Membership: |
|

ARHGAP1
Genomic Map