Assay Details
Gene Symbol: COA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930301.1 | - | 1 | 323 | 120 | |
BX934195.1 | - | 1 | 314 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: COA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930301.1 | - | 1 | 323 | 120 | |
BX934195.1 | - | 1 | 314 | 120 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |