Assay Details
| Gene Symbol: CDYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939667.3 | XP_004939724.1 | 7 | 2522 | 121 | |
| XM_015276006.1 | XP_015131492.1 | 8 | 2568 | 121 | ||
| XM_025147664.1 | XP_025003432.1 | 8 | 2651 | 121 | ||
| XM_025147665.1 | XP_025003433.1 | 8 | 2578 | 121 | ||
| XM_418964.6 | XP_418964.2 | 7 | 2801 | 121 | ||
| GenBank mRNA | CR406523.1 | - | 1 | 224 | 121 | |
More Information
Set Membership: |
|

CDYL
Genomic Map