Assay Details
| Gene Symbol: GNA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233236.5 | XP_001233237.2 | 4 | 7723 | 68 | |
| GenBank mRNA | CR406561.1 | - | 1 | 417 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNA12| Gene Symbol: GNA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233236.5 | XP_001233237.2 | 4 | 7723 | 68 | |
| GenBank mRNA | CR406561.1 | - | 1 | 417 | 68 | |
Set Membership: |
|