Assay Details
| Gene Symbol: HM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642489.4 | XP_003642537.3 | 3 - 4 | 400 | 59 | |
| XM_004947155.3 | XP_004947212.2 | 3 - 4 | 400 | 59 | ||
| GenBank mRNA | CR406652.1 | - | 3 - 4 | 266 | 59 | |
More Information
Set Membership: |
|

HM13
Genomic Map