Assay Details
| Gene Symbol: TOR1AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943216.2 | XP_004943273.1 | 5 | 2240 | 85 | |
| XM_015290501.2 | XP_015145987.1 | 6 | 2294 | 85 | ||
| XM_015290502.2 | XP_015145988.1 | 6 | 3443 | 85 | ||
| XM_422261.6 | XP_422261.3 | 6 | 2305 | 85 | ||
| GenBank mRNA | CR406811.1 | - | 1 | 1020 | 85 | |
More Information
Set Membership: |
|

TOR1AIP1
Genomic Map