Assay Details
| Gene Symbol: MED19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426411.5 | XP_426411.2 | 3 - 4 | 582 | 83 | |
| GenBank mRNA | CR407021.1 | - | 1 - 2 | 33 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED19| Gene Symbol: MED19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426411.5 | XP_426411.2 | 3 - 4 | 582 | 83 | |
| GenBank mRNA | CR407021.1 | - | 1 - 2 | 33 | 83 | |
Set Membership: |
|