Assay Details
| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297583.1 | XP_015153069.1 | 1 | 2156 | 75 | |
| GenBank mRNA | CR407050.1 | - | 1 | 722 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTR1D| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297583.1 | XP_015153069.1 | 1 | 2156 | 75 | |
| GenBank mRNA | CR407050.1 | - | 1 | 722 | 75 | |
Set Membership: |
|