Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298671.2 | XP_015154157.1 | 5 - 6 | 727 | 64 | |
| XM_015298672.2 | XP_015154158.1 | 5 - 6 | 727 | 64 | ||
| XM_015298673.2 | XP_015154159.1 | 5 - 6 | 727 | 64 | ||
| XM_015298674.2 | XP_015154160.1 | 5 - 6 | 727 | 64 | ||
| GenBank mRNA | CR407256.1 | - | 1 - 2 | 161 | 64 | |
More Information
Set Membership: |
|

CADM3
Genomic Map